Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ocln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031329.2 | NP_112619.2 | 4 | 973 | ||
XM_006231853.2 | XP_006231915.1 | 4 | 969 | |||
XM_006231854.2 | XP_006231916.1 | 4 | 1013 | |||
XM_006231855.2 | XP_006231917.1 | 3 | 893 | |||
GenBank mRNA | BC062037.1 | - | 4 | 973 | 5623744 |