Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031350.1 | NP_112640.1 | 4, 5 | 421 | ||
GenBank mRNA | AB035306.1 | - | 4, 5 | 421 | ||
BC062046.1 | - | 4, 5 | 512 | 5597999 |