Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gipc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053341.1 | NP_445793.1 | 2 | 379 | ||
XM_006255311.2 | XP_006255373.1 | 2 | 2644 | |||
XM_006255312.2 | XP_006255374.1 | 3 | 1638 | |||
XM_008772420.1 | XP_008770642.1 | 3 | 1568 | |||
GenBank mRNA | AF032120.1 | - | 2 | 379 | ||
AF089817.1 | - | 3 | 397 | |||
BC061964.1 | - | 2 | 1093 | |||
BC070505.1 | - | 3 | 422 | 7097697 |