Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chic2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105736.2 | NP_001099206.1 | 2 | 210 | ||
GenBank mRNA | BE109715.1 | - | 3 | 188 | ||
BE112096.1 | - | 2 | 210 | |||
EU202679.1 | - | 2 | 156 | |||
EU202680.1 | - | 2 | 156 |