Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032067.1 | NP_114456.1 | 5 | 1387 | ||
GenBank mRNA | BC089788.1 | - | 5 | 1441 | 7374343 | |
U82623.1 | - | 5 | 1387 |