Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053599.2 | NP_446051.2 | 3 | 495 | ||
XM_008761227.1 | XP_008759449.1 | 4 | 1079 | |||
XM_008761228.1 | XP_008759450.1 | 3 | 2027 | |||
GenBank mRNA | BC070514.1 | - | 3 | 495 | 7100462 | |
D38056.1 | - | 3 | 468 |