Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bcl2a1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007536.2 | NP_031562.1 | 2 | 620 | ||
GenBank mRNA | AK165576.1 | - | 2 | 372 | ||
BC027536.1 | - | 2 | 533 | 1363928 | ||
BC053965.1 | - | 2 | 642 | |||
BC116240.2 | - | 2 | 576 | 40091847 |
Gene Symbol: Bcl2a1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009742.3 | NP_033872.1 | 2 | 620 | ||
GenBank mRNA | BC028762.1 | - | 566 | 1226745 | ||
L16462.1 | - | 2 | 620 |