Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifna6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_206871.1 | NP_996754.1 | 1 | 235 | ||
GenBank mRNA | BC151087.1 | - | 1 | 375 | 9088000 |
Gene Symbol: Ifnab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008336.2 | NP_032362.2 | 1 | 276 | ||
GenBank mRNA | BC116862.1 | - | 1 | 276 | 40126181 | |
BC116864.1 | - | 1 | 276 | 40126183 |