Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPLA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007260.2 | NP_009191.1 | 7 | 429 | ||
XM_005245728.4 | XP_005245785.1 | 7 | 901 | |||
XM_017000165.1 | XP_016855654.1 | 7 | 796 | |||
GenBank mRNA | AF090423.1 | - | 7 | 475 | ||
AF098668.1 | - | 7 | 429 | |||
AK024724.1 | - | 3 | 109 | |||
BC017034.2 | - | 7 | 394 | 3845764 | ||
BC017193.2 | - | 7 | 383 | 3138786 |
Gene Symbol: LYPLA2P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033250.1 | 1 | 418 |