Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDIL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278642.1 | NP_001265571.1 | 4 | 855 | ||
NM_005711.4 | NP_005702.3 | 5 | 885 | |||
GenBank mRNA | AK312858.1 | - | 5 | 402 | ||
BC030828.1 | - | 5 | 811 | 4791845 | ||
BC053656.1 | - | 4 | 436 | 5753154 | ||
BX648583.1 | - | 4 | 374 | |||
U70312.1 | - | 5 | 502 | |||
U70313.1 | - | 4 | 362 |