Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005763.3 | NP_005754.2 | 9 | 1078 | ||
XM_011515725.2 | XP_011514027.1 | 9 | 1057 | |||
GenBank mRNA | AF229180.1 | - | 9 | 1002 | ||
AJ007714.2 | - | 8 | 1029 | |||
AK225898.1 | - | 9 | 971 | |||
BC144247.1 | - | 8 | 988 |