Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006327.3 | NP_006318.1 | 3 | 417 | ||
NR_073029.1 | 3 | 417 | ||||
NR_073030.1 | 2 | 358 | ||||
XM_011539098.1 | XP_011537400.1 | 3 | 355 | |||
XM_011539099.1 | XP_011537401.1 | 3 | 355 | |||
GenBank mRNA | AF030162.1 | - | 3 | 369 | ||
AK223331.1 | - | 3 | 357 | |||
AK304883.1 | - | 3 | 352 | |||
AY442341.1 | - | 3 | 369 | |||
BC062707.1 | - | 3 | 418 | 4772587 | ||
BC066951.1 | - | 3 | 367 | 5264814 | ||
BY799488.2 | - | 2 | 291 | |||
CD514420.1 | - | 3 | 293 |