Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIMM23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006327.3 | NP_006318.1 | 2 | 322 | ||
NR_073029.1 | 2 | 322 | ||||
XM_011539098.1 | XP_011537400.1 | 2 | 260 | |||
XM_011539099.1 | XP_011537401.1 | 2 | 260 | |||
GenBank mRNA | AF030162.1 | - | 2 | 274 | ||
AK223331.1 | - | 2 | 262 | |||
AK304883.1 | - | 2 | 257 | |||
AY442341.1 | - | 2 | 274 | |||
BC062707.1 | - | 2 | 326 | 4772587 | ||
BC066951.1 | - | 2 | 272 | 5264814 | ||
CD514420.1 | - | 2 | 198 |
Gene Symbol: TIMM23B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290117.1 | NP_001277046.1 | 2 | 312 | ||
NM_001290118.1 | NP_001277047.1 | 2 | 312 | |||
NR_110767.1 | 3 | 436 | ||||
NR_135475.1 | 2 | 312 | ||||
XM_011539104.1 | XP_011537406.1 | 2 | 264 | |||
XM_017015460.1 | XP_016870949.1 | 2 | 264 | |||
XM_017015461.1 | XP_016870950.1 | 2 | 264 | |||
XM_017015462.1 | XP_016870951.1 | 2 | 264 | |||
GenBank mRNA | AK098044.1 | - | 3 | 421 | ||
AK293228.1 | - | 2 | 264 | |||
AK295370.1 | - | 2 | 264 | |||
AK316490.1 | - | 2 | 264 | |||
AK316549.1 | - | 2 | 313 | |||
AL542628.3 | - | 3 | 423 | |||
DA019889.1 | - | 2 | 265 |