Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006349.2 | NP_006340.1 | 5 | 1164 | ||
XM_011515739.2 | XP_011514041.1 | 5 | 895 | |||
GenBank mRNA | AK125123.1 | - | 6 | 4161 | ||
BC017333.2 | - | 5 | 1164 | 4647446 | ||
U61837.1 | - | 5 | 702 |