Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGPAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012727.1 | NP_001012745.1 | 4, 5 | 662 | ||
NM_006412.3 | NP_006403.2 | 5, 6 | 758 | |||
GenBank mRNA | AF000237.1 | - | 5, 6 | 722 | ||
AF011374.1 | - | 5, 6 | 716 | |||
BC000026.2 | - | 5, 6 | 701 | 3507150 | ||
BC004529.2 | - | 4, 5 | 596 | 3347049 | ||
BC007269.1 | - | 3, 4 | 333 | |||
BC019292.1 | - | 5, 6 | 710 | 2823580 | ||
U56418.2 | - | 5, 6 | 695 |