Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NFAT5 SIRNA Assays ›
Gene Symbol:
NFAT5
Gene Name:
nuclear factor of activated T-cells 5, tonicity-responsive
Gene Aliases:
NF-AT5, NFATL1, NFATZ, OREBP, TONEBP
Chromosome Location:
Chr.16: 69565952 - 69704666 on Build GRCh38
UniGene ID:
Hs.371987
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s223152
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NFAT5
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113178.2
NP_001106649.1
13
4422
NM_006599.3
NP_006590.1
12
4371
NM_138713.3
NP_619727.2
13
4425
NM_138714.3
NP_619728.2
14
4487
NM_173214.2
NP_775321.1
13
4433
NM_173215.2
NP_775322.1
13
4433
XM_006721125.3
XP_006721188.1
12
5096
XM_011522817.2
XP_011521119.1
12
4368
XM_011522818.2
XP_011521120.1
15
4560
XM_011522819.2
XP_011521121.1
15
4142
XM_011522820.2
XP_011521122.1
12
3572
XM_011522822.2
XP_011521124.1
11
3520
XM_017022870.1
XP_016878359.1
12
4720
XM_017022871.1
XP_016878360.1
13
4444
XM_017022872.1
XP_016878361.1
12
4301
XM_017022873.1
XP_016878362.1
12
3910
GenBank mRNA
AB020634.1
-
12
4353
AF089824.1
-
13
4375
AF134870.1
-
10
3807
AF163836.1
-
11
3894
AF346509.1
-
14
5373
AJ005683.1
-
13
4049
AJ005684.1
-
13
4049
AJ243298.1
-
13
4192
AJ243299.1
-
13
4184
AK000312.1
-
1
1046
BC131509.1
-
13
4326
40083218
BC146765.1
-
12
4353
9007208
U80231.1
-
1
1066
Z97016.1
-
1
2015
Back To Top
More Information
Panther Classification:
Molecular Function -
Rel homology transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
cytokine production
transcription from RNA polymerase II promoter
response to osmotic stress
signal transduction
excretion
positive regulation of transcription from RNA polymerase II promoter
regulation of calcineurin-NFAT signaling cascade
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-65d5d77f49-2ddqc:80/100.66.79.182:80.
git-commit: a4c025c8ba47546866237b65de1f276a5fdbbe78
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.18.1-Offline