Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPS8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006710.4 | NP_006701.1 | 7 | 1065 | ||
NM_198189.2 | NP_937832.1 | 8 | 1130 | |||
GenBank mRNA | AK289891.1 | - | 8 | 664 | ||
AK313437.1 | - | 7 | 608 | |||
BC003090.1 | - | 7 | 589 | 3503287 | ||
BC036499.1 | - | 7 | 1199 | 5264527 | ||
BC080617.1 | - | 7 | 561 | 6252628 | ||
BG721393.1 | - | 8 | 669 | |||
CR456994.1 | - | 7 | 541 | |||
U51205.1 | - | 7 | 590 |