Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHKA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277.2 | NP_001268.2 | 7 | 1111 | ||
NM_212469.1 | NP_997634.1 | 6 | 1057 | |||
XM_017017147.1 | XP_016872636.1 | 6 | 995 | |||
XM_017017148.1 | XP_016872637.1 | 6 | 2467 | |||
GenBank mRNA | BC036471.2 | - | 6 | 935 | 5265994 | |
D10704.1 | - | 7 | 921 |