Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBA3D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080386.3 | NP_525125.2 | 4 | 854 | ||
GenBank mRNA | AK093116.1 | - | 4 | 768 | ||
AK292253.1 | - | 4 | 741 | |||
AW966211.1 | - | 1 | 87 | |||
BC057810.1 | - | 4 | 740 | 5268074 |
Gene Symbol: TUBA3E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207312.2 | NP_997195.1 | 4 | 762 | ||
GenBank mRNA | AA397983.1 | - | 1 | 40 | ||
AK131043.1 | - | 4 | 740 | |||
BC057811.1 | - | 4 | 762 | 5298532 |