Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007263.3 | NP_009194.2 | 1 | 124 | ||
NM_199442.1 | NP_955474.1 | 1 | 124 | |||
NM_199444.1 | NP_955476.1 | 1 | 124 | |||
XM_011527655.1 | XP_011525957.1 | 1 | 198 | |||
XM_011527656.1 | XP_011525958.1 | 1 | 198 | |||
GenBank mRNA | AF092438.1 | - | 1 | 119 | ||
AJ131182.1 | - | 1 | 124 | |||
AJ249366.1 | - | 1 | 97 | |||
AK222584.1 | - | 1 | 121 | |||
AK315281.1 | - | 1 | 121 | |||
AL136928.1 | - | 1 | 101 | |||
BC003155.1 | - | 1 | 96 | 2906127 | ||
BC007250.1 | - | 1 | 101 | 2989183 | ||
BC017285.2 | - | 1 | 87 | 4891380 | ||
BI520280.1 | - | 1 | 137 | |||
CR456886.1 | - | 1 | 82 |