Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303473.1 | NP_001290402.1 | 2 | 1036 | ||
NM_001303474.1 | NP_001290403.1 | 3 | 1304 | |||
NM_138445.3 | NP_612454.1 | 2 | 1159 | |||
XM_005249613.2 | XP_005249670.1 | 3 | 1713 | |||
XM_006715642.2 | XP_006715705.1 | 3 | 1881 | |||
XM_006715643.2 | XP_006715706.1 | 3 | 1831 | |||
XM_011515116.2 | XP_011513418.1 | 3 | 1278 | |||
GenBank mRNA | AK300166.1 | - | 2 | 742 | ||
AK313171.1 | - | 2 | 1125 | |||
BC014241.2 | - | 1 | 937 | 4563636 |
Set Membership: |
G-protein coupled receptor |