Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001015001.2 | NP_001015001.1 | 7, 8 | 1378 | ||
NM_001321926.1 | NP_001308855.1 | 6, 7 | 1246 | |||
NM_001321927.1 | NP_001308856.1 | 7, 8 | 1471 | |||
NM_001321928.1 | NP_001308857.1 | 7, 8 | 1232 | |||
NM_001321929.1 | NP_001308858.1 | 6, 7 | 1230 | |||
NR_135856.1 | 6, 7 | 1477 | ||||
XM_005254498.3 | XP_005254555.1 | 6, 7 | 953 | |||
XM_017022369.1 | XP_016877858.1 | 7, 8 | 1303 | |||
XM_017022370.1 | XP_016877859.1 | 7, 8 | 1228 | |||
GenBank mRNA | AK223365.1 | - | 6, 7 | 1058 | ||
AK295776.1 | - | 6, 7 | 1122 | |||
AK316124.1 | - | 7, 8 | 1329 | |||
AK316319.1 | - | 7, 8 | 1232 | |||
BC001926.1 | - | 6, 7 | 1063 | 2820642 | ||
BC006467.1 | - | 6, 7 | 989 | 3535486 | ||
BT006628.1 | - | 6, 7 | 886 |
Gene Symbol: CKMT1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020990.4 | NP_066270.1 | 7, 8 | 1475 | ||
NR_135748.1 | 7 | 2341 | ||||
NR_135749.1 | 7 | 2321 | ||||
NR_135750.1 | 8 | 2292 | ||||
NR_135751.1 | 7 | 2181 | ||||
NR_135752.1 | 6 | 1948 | ||||
XM_005254150.3 | XP_005254207.1 | 6, 7 | 953 | |||
XM_011521194.1 | XP_011519496.1 | 7, 8 | 1563 | |||
XM_011521195.2 | XP_011519497.1 | 7, 8 | 1302 | |||
XM_011521196.1 | XP_011519498.1 | 7, 8 | 1228 | |||
XM_011521197.2 | XP_011519499.1 | 6, 7 | 1062 | |||
XM_011521198.1 | XP_011519500.1 | 7, 8 | 1747 | |||
XM_011521199.2 | XP_011519501.2 | 7, 8 | 1519 | |||
XM_017021902.1 | XP_016877391.1 | 7, 8 | 1703 | |||
GenBank mRNA | AK094322.1 | - | 8 | 1870 | ||
BC108652.1 | - | 6, 7 | 998 | 6190035 | ||
BC121001.1 | - | 6, 7 | 1062 | 40116549 | ||
BC121002.1 | - | 6, 7 | 1062 | 40116552 |