Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLNS1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293.2 | NP_001284.1 | 5 | 614 | ||
NM_001311199.1 | NP_001298128.1 | 5 | 614 | |||
NM_001311200.1 | NP_001298129.1 | 4 | 512 | |||
NM_001311202.1 | NP_001298131.1 | 3 | 404 | |||
GenBank mRNA | AF005422.1 | - | 5 | 552 | ||
AF026003.1 | - | 5 | 522 | |||
AK315259.1 | - | 5 | 608 | |||
AL530926.3 | - | 4 | 458 | |||
BC119634.1 | - | 5 | 571 | 40115653 | ||
BC119635.1 | - | 5 | 571 | 40115655 | ||
BQ643786.1 | - | 5 | 584 | |||
BT019907.1 | - | 5 | 522 | |||
BU169662.1 | - | 3 | 392 | |||
U17899.1 | - | 5 | 596 | |||
U53454.1 | - | 5 | 606 | |||
X91788.1 | - | 5 | 610 |