Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRIG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153377.4 | NP_700356.2 | 1, 2 | 548 | ||
XM_017018790.1 | XP_016874279.1 | 1, 2 | 550 | |||
GenBank mRNA | AK292857.1 | - | 1, 2 | 529 | ||
AY358288.1 | - | 1, 2 | 491 | |||
AY505340.1 | - | 1, 2 | 299 | |||
BC126169.1 | - | 1, 2 | 444 | 8991885 | ||
BC126171.1 | - | 1, 2 | 444 | 8991887 | ||
BC143960.1 | - | 1, 2 | 444 |