Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152329.3 | NP_689542.2 | 4 | 1348 | ||
NM_203467.1 | NP_982292.1 | 3 | 626 | |||
NR_037792.1 | 6 | 1568 | ||||
NR_037793.1 | 5 | 1456 | ||||
GenBank mRNA | AK293156.1 | - | 4 | 1067 | ||
AY052405.1 | - | 4 | 1034 | |||
BC030142.2 | - | 3 | 562 | |||
BC064133.1 | - | 5 | 1201 | |||
BC093697.1 | - | 3 | 328 | |||
BC107888.1 | - | 3 | 375 | |||
BC112241.1 | - | 3 | 328 | |||
BC139921.1 | - | 4 | 1088 | 40148567 | ||
BQ059807.1 | - | 4 | 363 | |||
BX248298.1 | - | 4 | 1091 | |||
BX648029.1 | - | 6 | 1491 |