Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300LB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174892.3 | NP_777552.3 | 2 | 419 | ||
XM_005257027.3 | XP_005257084.1 | 2 | 419 | |||
GenBank mRNA | AF427618.1 | - | 2 | 319 | ||
AK290925.1 | - | 2 | 419 | |||
AK298665.1 | - | 2 | 420 | |||
AY359025.1 | - | 2 | 348 | |||
AY646929.1 | - | 2 | 366 | |||
BC028091.1 | - | 2 | 411 | 5219239 | ||
DA673717.1 | - | 2 | 421 |