Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PODN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153703.4 | NP_714914.2 | 9 | 1848 | ||
XM_005270457.3 | XP_005270514.1 | 10 | 1877 | |||
XM_011540677.2 | XP_011538979.1 | 5 | 1002 | |||
XM_017000294.1 | XP_016855783.1 | 9 | 2198 | |||
GenBank mRNA | AK296649.1 | - | 10 | 1743 | ||
AK299375.1 | - | 11 | 1933 | |||
AK300847.1 | - | 4 | 788 | |||
AK304599.1 | - | 5 | 1261 | |||
AK314096.1 | - | 9 | 1442 | |||
AY313607.1 | - | 9 | 1687 | |||
AY358294.1 | - | 11 | 1964 | |||
BC028292.1 | - | 9 | 1671 | 4938380 | ||
BC030608.2 | - | 9 | 1689 | 4826932 | ||
BQ719232.1 | - | 2 | 510 |