Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256129.1 | NP_001243058.1 | 1, 2 | 292 | ||
NM_138795.3 | NP_620150.1 | 1, 2 | 292 | |||
GenBank mRNA | AB118752.1 | - | 1, 2 | 121 | ||
AK127138.1 | - | 1, 2 | 245 | |||
AK313122.1 | - | 1, 2 | 270 | |||
BC015408.1 | - | 1, 2 | 223 | 4391821 | ||
BG876867.1 | - | 1, 2 | 133 | |||
BM548962.1 | - | 1, 2 | 277 | |||
BP325806.1 | - | 1, 2 | 292 |