Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEU4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167599.2 | NP_001161071.1 | 3 | 885 | ||
NM_001167600.2 | NP_001161072.1 | 3 | 562 | |||
NM_001167601.2 | NP_001161073.1 | 3 | 565 | |||
NM_001167602.2 | NP_001161074.1 | 4 | 1063 | |||
NM_080741.3 | NP_542779.2 | 3 | 882 | |||
GenBank mRNA | AJ277883.1 | - | 2 | 349 | ||
AK025617.1 | - | 3 | 434 | |||
AK091038.1 | - | 4 | 1060 | |||
AK096992.1 | - | 2 | 643 | |||
AK289421.1 | - | 3 | 439 | |||
AK290076.1 | - | 3 | 422 | |||
AK298403.1 | - | 4 | 394 | |||
AK300969.1 | - | 3 | 435 | |||
AY203952.1 | - | 3 | 418 | |||
BC012899.1 | - | 3 | 836 | 4156395 | ||
BC105974.1 | - | 3 | 398 | 4944243 | ||
BC117214.1 | - | 3 | 526 | 40125765 |