Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173077.2 | NP_775100.1 | 9 | 987 | ||
XM_017003372.1 | XP_016858861.1 | 10 | 7094 | |||
GenBank mRNA | AJ422118.1 | - | 9 | 941 | ||
BC112076.1 | - | 9 | 987 | 8327544 | ||
BC112078.1 | - | 9 | 987 | 8327546 |