Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007372.3 | NP_031398.2 | 11, 12 | 1434 | ||
NM_203499.2 | NP_987095.1 | 10, 11 | 1363 | |||
XM_006721657.1 | XP_006721720.1 | 11, 12 | 1571 | |||
XM_011524259.2 | XP_011522561.1 | 5, 6 | 1372 | |||
XM_017024111.1 | XP_016879600.1 | 12, 13 | 1670 | |||
XM_017024112.1 | XP_016879601.1 | 6, 7 | 750 | |||
GenBank mRNA | AB036090.1 | - | 9, 10 | 1159 | ||
AF083255.1 | - | 8, 9 | 819 | |||
AK095772.1 | - | 8, 9 | 848 | |||
AK126480.1 | - | 3, 4 | 1461 | |||
AK292908.1 | - | 8, 9 | 946 | |||
BC015505.1 | - | 9, 10 | 1144 | 3905607 | ||
BC078667.1 | - | 9, 10 | 1102 | |||
BC093081.1 | - | 11, 12 | 1398 | 30407786 |