Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308056.1 | NP_001294985.1 | 5 | 562 | ||
NM_001308057.1 | NP_001294986.1 | 5 | 568 | |||
NM_001308058.1 | NP_001294987.1 | 5 | 562 | |||
NM_001308059.1 | NP_001294988.1 | 6 | 597 | |||
NM_001308060.1 | NP_001294989.1 | 3 | 327 | |||
NM_001308061.1 | NP_001294990.1 | 4 | 454 | |||
NM_139164.2 | NP_631903.1 | 5 | 568 | |||
NR_131752.1 | 4 | 454 | ||||
NR_131753.1 | 4 | 441 | ||||
XM_017009041.1 | XP_016864530.1 | 4 | 379 | |||
XM_017009042.1 | XP_016864531.1 | 4 | 373 | |||
XM_017009043.1 | XP_016864532.1 | 3 | 291 | |||
XM_017009044.1 | XP_016864533.1 | 4 | 361 | |||
GenBank mRNA | AF480299.1 | - | 4 | 346 | ||
AK054566.1 | - | 5 | 430 | |||
AK290312.1 | - | 5 | 481 | |||
AK308851.1 | - | 4 | 392 | |||
AK310848.1 | - | 4 | 365 | |||
AK315863.1 | - | 3 | 265 | |||
AK315869.1 | - | 4 | 379 | |||
AL710253.1 | - | 5 | 527 | |||
AL832599.2 | - | 1 | 24 | |||
BC019632.1 | - | 4 | 367 | |||
BC042956.1 | - | 5 | 566 | 5278105 | ||
BP234859.1 | - | 6 | 512 | |||
BP247836.1 | - | 4 | 371 | |||
DT220890.1 | - | 5 | 499 |