Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX6C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004374.3 | NP_004365.1 | 4 | 747 | ||
XM_017013020.1 | XP_016868509.1 | 4 | 553 | |||
GenBank mRNA | BC000187.2 | - | 3 | 372 | 3350637 | |
S82616.1 | - | 2 | 222 | |||
X13238.1 | - | 4 | 406 |