Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286822.1 | NP_001273751.1 | 3 | 1165 | ||
NM_004339.3 | NP_004330.1 | 6 | 1493 | |||
NR_104597.1 | 5 | 1384 | ||||
GenBank mRNA | AF149785.1 | - | 6 | 1430 | ||
AK094882.1 | - | 7 | 1198 | |||
AK095586.1 | - | 1 | 1794 | |||
AK293318.1 | - | 3 | 894 | |||
AK298557.1 | - | 3 | 953 | |||
BC000415.2 | - | 6 | 1251 | 2961420 | ||
BC012858.1 | - | 6 | 1276 | 3857505 | ||
BC019295.1 | - | 6 | 1240 | 2823814 | ||
BC020983.1 | - | 6 | 1257 | 3851447 | ||
BC031097.1 | - | 6 | 1271 | 5288388 | ||
BC034250.1 | - | 6 | 1265 | 3862830 | ||
L48984.1 | - | 3 | 891 | |||
Z50022.1 | - | 6 | 1297 |