Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007372.3 | NP_031398.2 | 15 | 1820 | ||
NM_203499.2 | NP_987095.1 | 14 | 1749 | |||
XM_006721657.1 | XP_006721720.1 | 15 | 1957 | |||
XM_011524259.2 | XP_011522561.1 | 9 | 1758 | |||
XM_017024111.1 | XP_016879600.1 | 16 | 2056 | |||
XM_017024112.1 | XP_016879601.1 | 10 | 1136 | |||
GenBank mRNA | AB036090.1 | - | 13 | 1545 | ||
AF083255.1 | - | 12 | 1205 | |||
AK001964.1 | - | 1 | 30 | |||
AK095772.1 | - | 12 | 1234 | |||
AK122737.1 | - | 1 | 799 | |||
AK126480.1 | - | 7 | 1847 | |||
AK292908.1 | - | 12 | 1332 | |||
BC015505.1 | - | 13 | 1530 | 3905607 | ||
BC078667.1 | - | 13 | 1488 | |||
BC093081.1 | - | 15 | 1784 | 30407786 |