Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAAR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175057.3 | NP_778227.3 | 1 | 847 | ||
GenBank mRNA | AY183469.1 | - | 1 | 845 | ||
BC069083.1 | - | 1 | 847 |
Set Membership: |
G-protein coupled receptor |