Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COX7C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001867.2 | NP_001858.1 | 3 | 356 | ||
GenBank mRNA | AK026505.1 | - | 3 | 382 | ||
BC001005.2 | - | 3 | 356 | 2821167 | ||
BC007498.2 | - | 3 | 320 | 3030363 | ||
X16560.1 | - | 3 | 285 |