Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178469.3 | NP_848564.2 | 4, 5 | 676 | ||
XM_017014272.1 | XP_016869761.1 | 4, 5 | 404 | |||
GenBank mRNA | AK126902.1 | - | 3, 4 | 2229 | ||
AY301268.1 | - | 4, 5 | 396 | |||
BC130465.1 | - | 4, 5 | 631 | 40146496 | ||
BC132714.1 | - | 4, 5 | 631 | 40146736 |