Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTRB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025200.3 | NP_001020371.3 | 6 | 616 | ||
GenBank mRNA | AK124403.1 | - | 3 | 2015 | ||
AK131056.1 | - | 6 | 603 | |||
AK225933.1 | - | 6 | 603 | |||
AK291822.1 | - | 6 | 601 | |||
BC073145.1 | - | 6 | 592 | 5747536 |
Gene Symbol: CTRB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC005385.1 | - | 6 | 625 | 3950220 | |
BT007356.1 | - | 5 | 583 | |||
CA946996.1 | - | 5 | 544 | |||
M24400.1 | - | 5 | 599 |