Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001916.4 | NP_001907.2 | 3 | 492 | ||
XM_017013102.1 | XP_016868591.1 | 2 | 1025 | |||
GenBank mRNA | AK026633.1 | - | 3 | 450 | ||
AK225796.1 | - | 2 | 1093 | |||
BC001006.1 | - | 3 | 438 | 2821836 | ||
BC015616.2 | - | 3 | 432 | 4840922 | ||
BC020566.1 | - | 3 | 431 | 4157475 | ||
BC024650.1 | - | 3 | 399 | |||
BG328536.1 | - | 3 | 492 | |||
BT019798.1 | - | 3 | 427 | |||
CN369438.1 | - | 3 | 434 | |||
CR541674.1 | - | 3 | 427 | |||
CR735564.1 | - | 3 | 508 | |||
M16597.1 | - | 2 | 133 | |||
X06994.1 | - | 3 | 435 |