Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001349.3 | NP_001340.2 | 2 | 297 | ||
XM_017003489.1 | XP_016858978.1 | 2 | 268 | |||
GenBank mRNA | AK129521.1 | - | 2 | 239 | ||
AK222476.1 | - | 2 | 127 | |||
AK290607.1 | - | 2 | 154 | |||
AY762100.1 | - | 2 | 116 | |||
BC000629.2 | - | 2 | 130 | 3344322 | ||
BC107749.1 | - | 2 | 110 | 6709590 | ||
BI753718.1 | - | 2 | 270 | |||
BT006710.1 | - | 2 | 81 | |||
DA597505.1 | - | 2 | 297 | |||
J05032.1 | - | 2 | 171 |