Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138328.2 | NP_612201.1 | 6 | 768 | ||
XM_006721733.3 | XP_006721796.1 | 5 | 745 | |||
XM_006721734.3 | XP_006721797.1 | 5 | 1137 | |||
XM_011524443.2 | XP_011522745.1 | 4 | 586 | |||
XM_017024272.1 | XP_016879761.1 | 6 | 773 | |||
XM_017024273.1 | XP_016879762.1 | 5 | 748 | |||
XM_017024274.1 | XP_016879763.1 | 5 | 1076 | |||
XM_017024275.1 | XP_016879764.1 | 5 | 614 | |||
XM_017024276.1 | XP_016879765.1 | 4 | 588 | |||
XM_017024277.1 | XP_016879766.1 | 5 | 677 | |||
XM_017024278.1 | XP_016879767.1 | 5 | 704 | |||
XM_017024279.1 | XP_016879768.1 | 4 | 1411 | |||
XM_017024280.1 | XP_016879769.1 | 3 | 728 | |||
GenBank mRNA | AJ313480.1 | - | 6 | 768 | ||
AK124570.1 | - | 4 | 582 | |||
BC100975.1 | - | 6 | 767 | 40005244 | ||
BC100976.1 | - | 6 | 768 | |||
BC100977.1 | - | 5 | 744 | 40005246 | ||
BC100978.1 | - | 6 | 747 | 40005247 |