Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080411.2 | NP_001073880.1 | 4 | 423 | ||
NM_001308346.1 | NP_001295275.1 | 5 | 482 | |||
NM_001308348.1 | NP_001295277.1 | 4 | 414 | |||
NM_001308351.1 | NP_001295280.1 | 6 | 520 | |||
NM_001308355.1 | NP_001295284.1 | 5 | 473 | |||
NM_001308357.1 | NP_001295286.1 | 5 | 461 | |||
XM_006722661.3 | XP_006722724.1 | 4 | 322 | |||
XM_011527725.2 | XP_011526027.1 | 4 | 384 | |||
XM_011527728.2 | XP_011526030.1 | 6 | 410 | |||
XM_011527729.2 | XP_011526031.1 | 5 | 351 | |||
XM_017026413.1 | XP_016881902.1 | 3 | 954 | |||
GenBank mRNA | AK098300.1 | - | 4 | 392 | ||
BC047412.1 | - | 5 | 396 | 5295132 | ||
CN345816.1 | - | 4 | 339 | |||
DB450226.1 | - | 5 | 408 | |||
DB454511.1 | - | 5 | 453 |