Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF540 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172225.2 | NP_001165696.1 | 5 | 1092 | ||
NM_001172226.2 | NP_001165697.1 | 4 | 996 | |||
NM_152606.4 | NP_689819.1 | 5 | 1121 | |||
GenBank mRNA | AK122597.1 | - | 5 | 1041 | ||
AK131388.1 | - | 1 | 1046 | |||
AK290486.1 | - | 5 | 1042 | |||
AL832100.1 | - | 4 | 997 | |||
AL832315.1 | - | 5 | 1086 | |||
AL832687.1 | - | 5 | 1045 | |||
BC017940.1 | - | 5 | 1046 | |||
BC126478.1 | - | 5 | 892 | 8992194 | ||
BC126480.1 | - | 5 | 892 | 8992196 | ||
BX537980.1 | - | 5 | 1037 |