Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTTG1IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286822.1 | NP_001273751.1 | 2 | 435 | ||
NM_004339.3 | NP_004330.1 | 2 | 435 | |||
NR_104597.1 | 2 | 435 | ||||
GenBank mRNA | AF149785.1 | - | 2 | 373 | ||
AK094882.1 | - | 2 | 168 | |||
AK290139.1 | - | 2 | 188 | |||
AK298557.1 | - | 2 | 223 | |||
AK315626.1 | - | 2 | 250 | |||
BC000415.2 | - | 2 | 192 | 2961420 | ||
BC012858.1 | - | 2 | 218 | 3857505 | ||
BC019295.1 | - | 2 | 182 | 2823814 | ||
BC020983.1 | - | 2 | 199 | 3851447 | ||
BC031097.1 | - | 2 | 213 | 5288388 | ||
BC034250.1 | - | 2 | 207 | 3862830 | ||
BG721942.1 | - | 2 | 374 | |||
CN295711.1 | - | 2 | 226 | |||
DC396020.1 | - | 2 | 436 | |||
Z50022.1 | - | 2 | 241 |