Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIGD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145715.2 | NP_663761.1 | 1 | 1291 | ||
GenBank mRNA | AK027653.2 | - | 1 | 1291 | ||
AL833679.1 | - | 1 | 382 | |||
EU794666.1 | - | 1 | 1291 |