Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CFD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317335.1 | NP_001304264.1 | 5 | 1093 | ||
NM_001928.3 | NP_001919.2 | 5 | 1072 | |||
GenBank mRNA | BI763909.1 | - | 448 | |||
M84526.1 | - | 5 | 1019 |