Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DFFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282669.1 | NP_001269598.1 | 6, 7 | 1185 | ||
NM_001320132.1 | NP_001307061.1 | 6, 7 | 1282 | |||
NM_001320136.1 | NP_001307065.1 | 6, 7 | 1163 | |||
NM_004402.3 | NP_004393.1 | 6, 7 | 1113 | |||
NR_104222.1 | 7, 8 | 1250 | ||||
NR_135151.1 | 7, 8 | 1200 | ||||
NR_135152.1 | 6, 7 | 1135 | ||||
XM_017000498.1 | XP_016855987.1 | 5, 6 | 1048 | |||
GenBank mRNA | AB013918.1 | - | 6, 7 | 776 | ||
AB028912.1 | - | 6, 7 | 1076 | |||
AB028913.1 | - | 7, 8 | 994 | |||
AF039210.1 | - | 6, 7 | 776 | |||
AF064019.1 | - | 6, 7 | 907 | |||
AF409060.1 | - | 8, 9 | 787 | |||
AF409061.1 | - | 6, 7 | 802 | |||
AF409062.1 | - | 6, 7 | 830 | |||
AK290877.1 | - | 6, 7 | 904 | |||
AK303065.1 | - | 6, 7 | 992 | |||
AY438267.1 | - | 6, 7 | 705 | |||
AY438268.1 | - | 3, 4 | 328 | |||
BC032827.1 | - | 7, 8 | 1062 | |||
BC048797.1 | - | 6, 7 | 1099 | 5204996 |