Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSBP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009998.3 | NP_001009998.1 | 17 | 1551 | ||
NM_032627.4 | NP_116016.1 | 18 | 1617 | |||
XM_005259790.3 | XP_005259847.1 | 18 | 1658 | |||
XM_006722665.3 | XP_006722728.1 | 18 | 1779 | |||
XM_006722666.2 | XP_006722729.1 | 18 | 1737 | |||
XM_006722668.2 | XP_006722731.1 | 17 | 1671 | |||
XM_017026431.1 | XP_016881920.1 | 17 | 2075 | |||
XM_017026432.1 | XP_016881921.1 | 17 | 2613 | |||
XM_017026433.1 | XP_016881922.1 | 17 | 2021 | |||
XM_017026434.1 | XP_016881923.1 | 16 | 2004 | |||
XM_017026435.1 | XP_016881924.1 | 16 | 2546 | |||
XM_017026436.1 | XP_016881925.1 | 16 | 1955 | |||
XM_017026437.1 | XP_016881926.1 | 17 | 1587 | |||
GenBank mRNA | AK098429.1 | - | 12 | 1132 | ||
BC000274.1 | - | 18 | 1524 | 3356283 |