Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLEC4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130441.2 | NP_569708.1 | 7 | 1093 | ||
NM_203503.1 | NP_987099.1 | 6 | 1000 | |||
XM_017018940.1 | XP_016874429.1 | 6 | 748 | |||
GenBank mRNA | AF293615.1 | - | 7 | 1093 | ||
AF325459.1 | - | 6 | 628 | |||
AF325460.1 | - | 5 | 516 | |||
AK122596.1 | - | 6 | 776 | |||
AY358223.1 | - | 6 | 759 | |||
BC074967.2 | - | 6 | 646 | 30915289 | ||
BC074968.2 | - | 6 | 646 | 30915508 | ||
BC102015.1 | - | 6 | 703 | 40030336 | ||
BC102016.1 | - | 6 | 703 | 40030338 | ||
BC102017.1 | - | 6 | 702 | 40030339 | ||
BC114338.1 | - | 6 | 703 | 40030323 |